EMAIL THIS PAGE TO A FRIEND
693758 Sigma-Aldrich

Neodymium

foil, thickness 1 mm, 99.9% (REM)

Purchase

Properties

assay   99.9% (REM)
form   foil
reaction suitability   reagent type: catalyst
core: neodymium
resistivity   64.0 μΩ-cm, 20°C
thickness   1 mm
bp   3074 °C (lit.)
mp   1021 °C (lit.)
density   7.003 g/mL at 25 °C (lit.)
SMILES string   [Nd]
InChI   1S/Nd
InChI key   QEFYFXOXNSNQGX-UHFFFAOYSA-N

Description

Application

Used in Physical Vapor Deposition (PVD) processes, including thermal and electron-beam (e-beam) evaporation, for the preparation of thin films.

Quantity

1 ea = 25 x 25 mm

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3178 4.1 / PGIII
WGK Germany 
3
RTECS 
QO8575000
Protocols & Articles

Articles

Functional Magnetic Materials: Synergy Between Basic Science and Evolving Technology

S. B. Roy Materials and Advanced Accelerator Sciences Division Raja Ramanna Centre for Advanced Technology Indore-452013, India Email: sbroy@rrcat.gov.in
Keywords: Crystallization, Microscopy, Phase transitions, Reductions, transformation

Metal Hydrides for NiMH Battery Applications

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications. The Partnership for a New Generation of V...
Dhanesh Chandra,* Wen-Ming Chien and Anjali Talekar
Material Matters Volume 6 Article 2
Keywords: Absorption, Automotive, Capillary electrophoresis, Combustion, Degradations, Electrochemical analysis, Electronics, Environmental, Ion-exchange chromatography, Phase transitions, Precipitation, Purification, Separation, Substitutions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?