• USA Home
  • 694258 - 5-Bromo-2-hexylthiophene

EMAIL THIS PAGE TO A FRIEND
694258 Sigma-Aldrich

5-Bromo-2-hexylthiophene

97%

Purchase

Properties

Related Categories Building Blocks, C10 to C20+, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks,
Quality Level   100
assay   97%
refractive index   n20/D 1.529
density   1.231 g/mL at 25 °C
SMILES string   CCCCCCc1ccc(Br)s1
InChI   1S/C10H15BrS/c1-2-3-4-5-6-9-7-8-10(11)12-9/h7-8H,2-6H2,1H3
InChI key   LSIYDKGTGHFTFW-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2810 6.1 / PGIII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Oligothiophene Synthesis

Oligothiophenes and their functional derivatives are used in a number of high-technology applications including OLEDs, OFETs and OPVs, where the ability to functionalize the oligothiophene backbone a...
Keywords: Building blocks, Coupling reactions, Sublimation, Suzuki coupling

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?