• USA Home
  • 698326 - 3-[(4′-Chloro-1-naphthyloxy)methyl]phenylboronic acid

EMAIL THIS PAGE TO A FRIEND
698326 Sigma-Aldrich

3-[(4′-Chloro-1-naphthyloxy)methyl]phenylboronic acid

90%

  • CAS Number 1257648-77-3

  • Empirical Formula (Hill Notation) C17H14BClO3

  • Molecular Weight 312.56

  •  PubChem Substance ID 329762002

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Monosubstituted Aryl Boronic Acids,
InChI Key   GWHYATBGYANRCV-UHFFFAOYSA-N
assay   90%
mp   181-204 °C

Description

Packaging

1, 5 g in glass bottle

Application

May contain varying amounts of anhydride

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?