• USA Home
  • 698377 - 1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
698377 Sigma-Aldrich

1,2-Bis[(2R,5R)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Synonym: (R,R)-Et-DUPHOS-Rh, [(R,R)Ethyl-DuPhosRh(COD)]BF4

  • CAS Number 228121-39-9

  • Empirical Formula (Hill Notation) C30H48BF4P2Rh

  • Molecular Weight 660.36

  •  PubChem Substance ID 329762007

  •  NACRES NA.22

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, DuPhos/BPE, Privileged Ligands and Complexes More...
Quality Level   100
SMILES string   [Rh+].F[B-](F)(F)F.C1CC=CCCC=C1.CC[C@@H]2CC[C@@H](CC)P2c3ccccc3P4[C@H](CC)CC[C@H]4CC
InChI   1S/C22H36P2.C8H12.BF4.Rh/c1-5-17-13-14-18(6-2)23(17)21-11-9-10-12-22(21)24-19(7-3)15-16-20(24)8-4;1-2-4-6-8-7-5-3-1;2-1(3,4)5;/h9-12,17-20H,5-8,13-16H2,1-4H3;1-2,7-8H,3-6H2;;/q;;-1;+1/b;2-1-,8-7-;;/t17-,18-,19-,20-;;;/m1.../s1
InChI key   DSYBEQKPSKLNMC-KYOOHHHUSA-N

Description

Application

DuPhos and BPE Ligands: Highly Efficient Privileged Ligands

Catalyst for:
• Asymmetric hydrogenation reactions

Packaging

50, 250 mg in amber glass bottle

Legal Information

Sold in collaboration with Kanata Chemical Technologies Inc. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

DuPhos and BPE Phospholane Ligands and Complexes

In the early 1990s, Burk and coworkers developed new electron-rich C2 symmetric bis(phospholane) ligands. The modular nature of these ligands allowed for variation of both phosphane substituent and b...
William Sommer and Daniel Weibel
ChemFiles 2008, 8.2, 34.
Keywords: Aminations, Asymmetric catalysis, Asymmetric synthesis, Building blocks, Catalysis, Crotylborations, Hydrogenations, Ligands, Reductions, Reductive aminations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?