• USA Home
  • 698660 - (R)-2-[(Boc-amino)methyl]-3-phenylpropionic acid

EMAIL THIS PAGE TO A FRIEND
698660 Aldrich

(R)-2-[(Boc-amino)methyl]-3-phenylpropionic acid

97%

Synonym: (R)-2-Benzyl-Boc-β-alanine, (R)-2-Benzyl-3-(Boc-amino)propionic acid

  • CAS Number 262301-38-2

  • Empirical Formula (Hill Notation) C15H21NO4

  • Molecular Weight 279.33

  •  MDL number MFCD07372496

Purchase

Properties

assay   97%
optical activity   [α]22/D +15.0°, c = 1 in chloroform
mp   94-102 °C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?