• USA Home
  • 698814 - Dicyclohexyl-(2,6-diisopropylphenyl)phosphine

EMAIL THIS PAGE TO A FRIEND
698814 Sigma-Aldrich

Dicyclohexyl-(2,6-diisopropylphenyl)phosphine

97%

Synonym: [2,6-Bis(1-methylethyl)phenyl]dicyclohexylphosphine

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
assay   97%
mp   96-100 °C
functional group   phosphine
reaction suitability   reaction type: Buchwald-Hartwig Cross Coupling Reaction
  reaction type: Heck Reaction
  reaction type: Hiyama Coupling
  reaction type: Negishi Coupling
  reaction type: Sonogashira Coupling
  reaction type: Stille Coupling
  reaction type: Suzuki-Miyaura Coupling
  reagent type: ligand
SMILES string   CC(C)c1cccc(C(C)C)c1P(C2CCCCC2)C3CCCCC3
InChI   1S/C24H39P/c1-18(2)22-16-11-17-23(19(3)4)24(22)25(20-12-7-5-8-13-20)21-14-9-6-10-15-21/h11,16-21H,5-10,12-15H2,1-4H3
InChI key   BTJYWUVKNAFRBF-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

250 mg in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?