• USA Home
  • 699659 - Umicore Metal Precursors Kit

EMAIL THIS PAGE TO A FRIEND
699659 Sigma-Aldrich

Umicore Metal Precursors Kit

  •  NACRES NA.22

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Homogeneous Pd Catalysts, Iridium Catalysts, Palladium Catalysts,
reaction suitability   reagent type: catalyst
core: iridium
  reagent type: catalyst
core: palladium
  reagent type: catalyst
core: rhodium
  reagent type: catalyst
core: ruthenium
storage temp.   −20°C

Description

Legal Information

A product of Umicore

Kit component also available separately

Description

Product #

Add to Cart

Chiralyst P1271, Umicore 500 mg SDS 693774
[Ir(cod)(acac)], Umicore 500 mg SDS 683086
HS157, Umicore, 97% 500 mg SDS 683094
Chiralyst P310, Umicore, 99% 500 mg SDS 683108
Chiralyst P294, Umicore 500 mg SDS 683116
Chiralyst P493, Umicore, 98% 500 mg SDS 683132
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate hydrate 500 mg SDS 683140
Chiralyst P468, Umicore 500 mg SDS 683159
Chiralyst P461, Umicore 500 mg SDS 683167
Chiralyst P374, Umicore 500 mg SDS 683175
Chiralyst P618, Umicore 500 mg SDS 686891
CARAC, Umicore, 98% 500 mg SDS 683183
Chiralyst P280, Umicore, 95% 500 mg SDS 683191
Chiralyst P978, Umicore 500 mg SDS 683205
Chiralyst P612, Umicore, 97% 500 mg SDS 683213
[Ru(benzene)Cl2]2, Umicore 500 mg SDS 683221
[Pd(allyl)Cl]2, Umicore 500 mg SDS 683396
[Pd(acac)2], Umicore, 99% 500 mg SDS 683388
[Pd(OAc)2]3, Umicore, 46.6-49.5% palladium (Pd) basis 500 mg SDS 683124
[Pd2(dba)3] x dba, Umicore 500 mg SDS 683345
Safety & Documentation

Safety Information

RIDADR 
UN 3316 9/PG 2
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?