• USA Home
  • 699772 - Polystyrene, azide terminated

EMAIL THIS PAGE TO A FRIEND
699772 Aldrich

Polystyrene, azide terminated

Mn 2,500

Synonym: Azide terminated polystyrene, PS N3 terminated, PS-N3, clickable PS

Purchase

Properties

Related Categories Hydrophobic Polymers, Materials Science, Polymer Science, Polymers, Polystyrene,
mol wt   Mn 2,500
mp   77-82 °C
Mw/Mn   <1.3

Description

Application

This is a well-defined polystyrene with an azide end group functionality. Azido groups are commonly used in Cu-mediated ligation (azide-alkyne click chemistry).

Legal Information

Sold for research purposes only and subject to field restriction: see www.sigmaaldrich.com/raftlicense. Patents: 7; 714; 075; 7; 250; 479; 7; 666; 962; 6; 642; 318; 6; 747; 111; W2010/8356.

Price and Availability


Webinar: Recent Developments in RAFT Agents
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?