• USA Home
  • 699780 - Poly(thiophene-3-[2-(2-methoxyethoxy)ethoxy]-2,5-diyl), sulfonated solution

EMAIL THIS PAGE TO A FRIEND
699780 Sigma-Aldrich

Poly(thiophene-3-[2-(2-methoxyethoxy)ethoxy]-2,5-diyl), sulfonated solution

2% in ethylene glycol monobutyl ether/water, 3:2, electronic grade

Synonym: Plexcore® OC RG-1200, Sulfonated polythiophene ink

  • CAS Number 1003582-37-3

  • Linear Formula (C9H12O6S2)X(C9H11O3S)Y

Purchase

Description

Application

Organic conductive ink suitable for spin coating applications and as hole injection layer in organic light emitting diodes (OLED).

Polmer-based Materials for Printed Electronics: Enabling High Efficency Solar Power and Lighting

Packaging

25 mL in glass bottle

Legal Information

Product of Plextronics, Inc. U.S. Patent number 6,166,172.

Plexcore is a registered trademark of Plextronics, Inc.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
194 °F
Flash Point(C) 
90 °C
Protocols & Articles

Articles

3D Printing of Carbon Fiber-Reinforced Composites

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries, most typically using a layer-by-layer fabrication process. 3D pri...
Zhenyu Bo* (Ph.D Candidate at Northwestern University) and Jia Choi*, PhD, Product Manager


*Materials Science Product Management Team, MilliporeSigma, Milwaukee, WI.
Keywords: Deposition, Nanomaterials, Nanotubes

Graphene-Based Transparent Conductive Electrodes

Kehan Yu1 and Junhong Chen2* 1Department of Chemical Engineering Case Western Reserve University Cleveland, Ohio 44106 2Department of Mechanical Engineering University of Wisconsin-Milwaukee, Milwauk...
Keywords: Catalysis, Centrifugation, Chemical vapor deposition, Deposition, Electronics, Microelectronics, Nanomaterials, Nanotubes, Oxidations, PAGE, Positron Emission Tomography, Purification, Raman spectroscopy, Reductions, Semiconductor, Solar cells, Solvents, Sonication, Spectroscopy

Inkjet Printing as a Key Enabling Technology for Printed Electronics

In the past decade, the family of digital printing technologies has evolved from being just a tool to visualize information into a generator of functionalities. The phrase ′printing beyond color′ apt...
Ashok Sridhar1, Thomas Blaudeck, Reinhard R. Baumann1,2*
Material Matters Volume 6 Article 1
Keywords: Absorption, Adhesion, Applications, Degradations, Deposition, Electronics, Industries, Microwave synthesis, Nanotechnology, Organic electronics, Printed electronics, Reproduction, Solar cells, Solvents, Spin coating, Type, Usage, transformation

Materials Design Concepts for Efficient Blue OLEDs:
A Joint Theoretical and Experimental Study

Since their discovery,1 organic light emitting devices (OLEDs) have evolved from a scientific curiosity into a technology with applications in flat panel displays and the potential to revolutionize t...
Evgueni Polikarpov, Asanga B. Padmaperuma
Keywords: Applications, Building blocks, Help, Materials Science, Methods, Purification, Reductions, Search, Support, Tools, Type

Nanoparticle-based Zinc Oxide Electron Transport Layers for Printed Organic Photodetectors

Gerardo Hernandez-Sosa,1,2* Ralph Eckstein,1,2 Tobias Rödlmeier,1,2 Uli Lemmer1,2,3 1Lichttechnisches Institut, Karlsruher Institut für Technologie, Engesserstrasse 13, 76131 Karlsruhe, Germany 2Inno...
Gerardo Hernandez-Sosa*, Ralph Eckstein, Tobias Rödlmeier, Uli Lemmer
Material Matters, 2016, 11.2
Keywords: Deposition, Electronics, Positron Emission Tomography, Recombination, Reductions, Solar cells, Thin film deposition

Plexcore® Organic Conductive Inks

Employing the regioselective polymerization techniques used to make P3HTs, Plextronics has developed a self-doping polymer poly(thiophene-3-[2[(2-methoxyethoxy)ethoxy]-2,5-diyl) (Figure 1), which is ...
Keywords: Degradations, Infrared spectroscopy, Polymerization reactions

Polymer-based Materials for Printed Electronics: Enabling High Efficiency Solar Power and Lighting

The soaring global demand for energy has created an urgent need for new energy sources that are both cost-competitive and eco-friendly. Renewable energy technology, such as solar power and energy eff...
Ritesh Tipnis*, Darin Laird, Mathew Mathai
Material Matters 2008, 3.4, 92.
Keywords: Absorption, Degradations, Deposition, Electronics, Environmental, Infrared spectroscopy, Organic electronics, PAGE, Photovoltaics, Polymerization reactions, Printed electronics, Recombination, Reductions, Renewable energy, Semiconductor, Separation, Solar cells, Spin coating

Protocols

Properties and Application of Poly(thiophene-3-[2-(2-methoxyethoxy)ethoxy]-2,5-diyl), sulfonated solutions

Poly(thiophene-3-[2-(2-methoxyethoxy)ethoxy]-2,5-diyl), sulfonated solutions are organic conductive inks designed for spin coating applications. However, they have also been evaluated and may be used...
Keywords: Deposition, Detection methods, Reductions, Semiconductor, Solvents, Spin coating

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?