• USA Home
  • 700215 - 2,2,6,6-Tetramethyl-3,5-heptanedionato lithium

EMAIL THIS PAGE TO A FRIEND
700215 Sigma-Aldrich

2,2,6,6-Tetramethyl-3,5-heptanedionato lithium

98%

Synonym: (Li(TMHD)), Lithium Dipivaloylmethanate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Lithium Catalysts More...
InChI Key   YGUDQHNOEYNYSP-CFYXSCKTSA-M
assay   98%
mp   275-278 °C

Description

Application

Reactant for synthesis of:
• Bis-cycloiridiated complexes for electroluminescent studies
• Cationic imido alkylidene complexes
• Polyaromatic ligands containing pyrene
• Mono-cyclometalated platinum complexes

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?