EMAIL THIS PAGE TO A FRIEND
700800 Sigma-Aldrich

(R)-ShiP

97%

Synonym: (11aR)-(+)-10,11,12,13-Tetrahydrodiindeno[7,1-de:1′,7′-fg][1,3,2]dioxaphosphocin-5-phenoxy, Phenyl-[(R)-1,1-spirobiindane-7,7-diyl]-phosphite

  • CAS Number 656233-53-3

  • Empirical Formula (Hill Notation) C23H19O3P

  • Molecular Weight 374.37

  •  MDL number MFCD08459341

Purchase

Properties

assay   97%
optical activity   [α]22/D +209°, c = 1 in chloroform
mp   91-105 °C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?