EMAIL THIS PAGE TO A FRIEND
70125 Aldrich

(−)-Myrtenal

≥97.0% (sum of enantiomers, GC)

Synonym: (1R)-2-Pinen-10-al, (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-en-2-carboxaldehyde

  • CAS Number 18486-69-6

  • Empirical Formula (Hill Notation) C10H14O

  • Molecular Weight 150.22

  •  Beilstein Registry Number 2961587

  •  MDL number MFCD00074768

  •  PubChem Substance ID 57652241

Purchase

Properties

Related Categories Aldehydes, Asymmetric Synthesis, Building Blocks, C10-C12, Carbonyl Compounds,
InChI Key   KMRMUZKLFIEVAO-IUCAKERBSA-N
assay   ≥97.0% (sum of enantiomers, GC)
optical activity   [α]20/D −14.6±1°, neat
refractive index   n20/D 1.504
density   0.988 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Terpene used as chiral building block, e.g. prep. of thromboxanes; Wittig and Wittig-Horner reactions; Transformation to pinocarveol

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Flash Point(F) 
172.4 °F
Flash Point(C) 
78 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?