• USA Home
  • 701939 - 2-Acetyl-3-thiopheneboronic acid

EMAIL THIS PAGE TO A FRIEND
701939 Sigma-Aldrich

2-Acetyl-3-thiopheneboronic acid

Synonym: 2-Acetyl-3-thienylboronic acid

Purchase

Properties

Related Categories Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Boronic Acids, Organometallic Reagents More...
mp   120-130 °C
storage temp.   2-8°C
SMILES string   CC(=O)c1sccc1B(O)O
InChI   1S/C6H7BO3S/c1-4(8)6-5(7(9)10)2-3-11-6/h2-3,9-10H,1H3
InChI key   QTGPYSQUEPGBHV-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Application

may contain varying amounts of anhydride

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?