• USA Home
  • 703060 - N-(Dicyclohexylphosphino)-2-(2′-methoxyphenyl)indole

EMAIL THIS PAGE TO A FRIEND
703060 Sigma-Aldrich

N-(Dicyclohexylphosphino)-2-(2′-methoxyphenyl)indole

97%

Synonym: 1-(Dicyclohexylphosphino)-2-(2′-methoxyphenyl)-1H-indole

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
assay   97%
mp   136-141 °C
functional group   phosphine
reaction suitability   reaction type: Buchwald-Hartwig Cross Coupling Reaction
  reaction type: Heck Reaction
  reaction type: Hiyama Coupling
  reaction type: Negishi Coupling
  reaction type: Sonogashira Coupling
  reaction type: Stille Coupling
  reaction type: Suzuki-Miyaura Coupling
  reagent type: ligand
SMILES string   COc1ccccc1-c2cc3ccccc3n2P(C4CCCCC4)C5CCCCC5
InChI   1S/C27H34NOP/c1-29-27-19-11-9-17-24(27)26-20-21-12-8-10-18-25(21)28(26)30(22-13-4-2-5-14-22)23-15-6-3-7-16-23/h8-12,17-20,22-23H,2-7,13-16H2,1H3
InChI key   CVYNBXIELBXSRJ-UHFFFAOYSA-N

Description

Application

Catalyst for Suzuki-Miyaura coupling

Packaging

1 g in glass bottle

250 mg in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?