• USA Home
  • 704717 - p-Terphenyl-4,4′′-dicarboxylic acid

EMAIL THIS PAGE TO A FRIEND
704717 Sigma-Aldrich

p-Terphenyl-4,4′′-dicarboxylic acid

Synonym: 1,4-Bis(4-carboxyphenyl)benzene

  • CAS Number 13653-84-4

  • Empirical Formula (Hill Notation) C20H14O4

  • Molecular Weight 318.32

  •  MDL number MFCD00389774

Purchase

Properties

mp   >500 °C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Metal Organic Frameworks

Metal-organic frameworks (MOFs) are materials in which metal – to-organic ligand interactions yield porous coordination networks with record-setting surface areas surpassing activated carbons and zeo...

Related Content

MOF Constructor

Metal-Organic Frameworks (MOFs) are a novel class of 3D networked, crystalline materials composed of metal ions or clusters connected by organic linkers. Given their porous nature and often high surf...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?