• USA Home
  • 705519 - 3-Chloro-6-(1-pyrrolidinyl)pyridazine

EMAIL THIS PAGE TO A FRIEND
705519 Sigma-Aldrich

3-Chloro-6-(1-pyrrolidinyl)pyridazine

97%

  • CAS Number 66346-85-8

  • Empirical Formula (Hill Notation) C8H10ClN3

  • Molecular Weight 183.64

  •  MDL number MFCD05864735

  •  PubChem Substance ID 329762567

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks, Pyridazines More...
Quality Level   100
assay   97%
mp   127-131 °C
storage temp.   2-8°C
SMILES string   Clc1ccc(nn1)N2CCCC2
InChI   1S/C8H10ClN3/c9-7-3-4-8(11-10-7)12-5-1-2-6-12/h3-4H,1-2,5-6H2
InChI key   NYOJAXJOBBCOEF-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Target organs 
Respiratory system
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?