• USA Home
  • 705640 - 1,7-Dimethyluric acid-2,4,5,6-13C4-1,3,9-15N3

EMAIL THIS PAGE TO A FRIEND
705640 Sigma-Aldrich

1,7-Dimethyluric acid-2,4,5,6-13C4-1,3,9-15N3

≥98 atom %, ≥98% (CP)

Synonym: 1,7-Dimethyl-2,6,8-trihydroxypurine--13C4,15N3 (2,4,5,6-13C4, 1,3,9-15N3)

  • CAS Number 1173023-17-0

  • Empirical Formula (Hill Notation) 13C4C3H815N3NO3

  • Molecular Weight 203.11

  •  MDL number MFCD11656117

  •  PubChem Substance ID 329762576

  •  NACRES NA.24

Purchase

Properties

Related Categories Caffeine and Caffeine Metabolites, Metabolites, Metabolites and Bioactive Compounds, Stable Isotopes
Quality Level   200
isotopic purity   ≥98 atom %
assay   ≥98% (CP)
mp   >300 °C
mass shift   M+7
storage temp.   −20°C
SMILES string   CN1C(=O)[15NH][13C]2=[13C]1[13C](=O)[15N](C)[13C](=O)[15NH]2
InChI   1S/C7H8N4O3/c1-10-3-4(8-6(10)13)9-7(14)11(2)5(3)12/h1-2H3,(H,8,13)(H,9,14)/i3+1,4+1,5+1,7+1,8+1,9+1,11+1
InChI key   NOFNCLGCUJJPKU-HNTYGULPSA-N

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?