• USA Home
  • 707082 - 1-Boc-4-iodopyrazole

EMAIL THIS PAGE TO A FRIEND
707082 Sigma-Aldrich

1-Boc-4-iodopyrazole

95%

Synonym: tert-Butyl 4-iodo-1H-pyrazole-1-carboxylate

Purchase

Properties

Related Categories Building Blocks, C3-C9, C7 to C8, Chemical Synthesis, Halogenated Heterocycles,
assay   95%
mp   67-71 °C
SMILES string   CC(C)(C)OC(=O)n1cc(I)cn1
InChI   1S/C8H11IN2O2/c1-8(2,3)13-7(12)11-5-6(9)4-10-11/h4-5H,1-3H3
InChI key   WRCRIGRVTPLDDD-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?