• USA Home
  • 707252 - Isopropenylboronic acid MIDA ester

EMAIL THIS PAGE TO A FRIEND
707252 Sigma-Aldrich

Isopropenylboronic acid MIDA ester

Purchase

Properties

Related Categories Alkenyl MIDA Boronates, Boronic Acids and Derivatives, Chemical Synthesis, MIDA Boronates, Organometallic Reagents More...
InChI Key   MZUQQYPADWTQLZ-UHFFFAOYSA-N
mp   136-140 °C

Description

Packaging

1 g in glass insert

5 g in glass bottle

Application

Reactant for:
• Stereoretentive Suzuki-Miyaura coupling

Suzuki Cross-Coupling with MIDA Boronates

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

MIDA Boronates for Suzuki–Miyaura Cross-Couplings

Professor Martin Burke and coworkers recently prepared retinal using key MIDA building block BB1 in iterative Suzuki-Miyaura cross coupling reactions (Scheme 1). The MIDA unit of BB1 is unreactive un...
Aaron Thornton
ChemFiles Volume 11 Article 1
Keywords: Building blocks, Coupling reactions, Cross couplings, Solvents

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?