• USA Home
  • 707392 - (1R,4R)-Bicyclo[2.2.2]octane-2,5-dione

EMAIL THIS PAGE TO A FRIEND
707392 Sigma-Aldrich

(1R,4R)-Bicyclo[2.2.2]octane-2,5-dione

95%

  • CAS Number 76478-53-0

  • Empirical Formula (Hill Notation) C8H10O2

  • Molecular Weight 138.16

  •  PubChem Substance ID 329762720

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Ketones, Organic Building Blocks More...
InChI Key   WMJKUNDVKHUCMV-PHDIDXHHSA-N
assay   95%
optical activity   [α]22/D −53.0°, c = 0.5 in chloroform
mp   202-206 °C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?