• USA Home
  • 707791 - 3-Chloro-4-pyridineboronic acid pinacol ester

EMAIL THIS PAGE TO A FRIEND
707791 Sigma-Aldrich

3-Chloro-4-pyridineboronic acid pinacol ester

97%

Purchase

Properties

Related Categories Boronate Esters, Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Boronate Esters, Organometallic Reagents More...
InChI Key   VUWRRLUCWJCPMF-UHFFFAOYSA-N
assay   97%
mp   86-90 °C

Description

Packaging

1 g in glass bottle

250 mg in glass insert

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Boronic Acid Esters - Chemfiles Volume 4 Article 2

The synthesis of biaryl compounds via the Suzuki–Miyaura coupling reaction has become more commonplace now that many arylboronic acids are readily available. Several years ago, Miyaura et al. demonst...
Chemfiles Volume 4 Article 2
Keywords: Coupling reactions, Suzuki-Miyaura coupling

Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?