EMAIL THIS PAGE TO A FRIEND
709212 Sigma-Aldrich

Gadolinium boride

powder, −325 mesh, 99.5% trace metals basis

Purchase

Properties

Related Categories 3D Printing Materials for Research and Development, Borides, Boron, Boron Catalysts, Catalysis and Inorganic Chemistry,
assay   99.5% trace metals basis
form   powder
particle size   −325 mesh
mp   2100 °C
density   5.27 g/cm3 at 25 °C

Description

Packaging

10 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

3D Printing of Carbon Fiber-Reinforced Composites

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries, most typically using a layer-by-layer fabrication process. 3D pri...
Zhenyu Bo* (Ph.D Candidate at Northwestern University) and Jia Choi*, PhD, Product Manager


*Materials Science Product Management Team, MilliporeSigma, Milwaukee, WI.
Keywords: Deposition, Nanomaterials, Nanotubes

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?