• USA Home
  • 710075 - (Thiomorpholinium-4-ylmethyl)trifluoroborate

EMAIL THIS PAGE TO A FRIEND
710075 Sigma-Aldrich

(Thiomorpholinium-4-ylmethyl)trifluoroborate

  • CAS Number 1268340-95-9

  • Empirical Formula (Hill Notation) C5H11BF3NS

  • Molecular Weight 185.02

  •  MDL number MFCD18427908

  •  PubChem Substance ID 329762533

  •  NACRES NA.22

Purchase

Properties

Related Categories Alkyl Trifluoroborate Salts, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents, Trifluoroborate Salts More...
Quality Level   100
description   internal salt
mp   146-156 °C
SMILES string   F[B-](F)(F)C[NH+]1CCSCC1
InChI   1S/C5H10BF3NS/c7-6(8,9)5-10-1-3-11-4-2-10/h1-5H2/q-1/p+1
InChI key   VUNVVPNMYVRSKZ-UHFFFAOYSA-O

Description

Application

Organotrifluoroborate involved in Suzuki-Miyaura cross-coupling reactions and cross coupling with mesylated phenol derivatives

Organotrifluoroborates as versatile and stable boronic acid surrogates

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?