• USA Home
  • 710970 - Dicyclohexyl(4-isopropylphenyl)phosphine

EMAIL THIS PAGE TO A FRIEND
710970 Sigma-Aldrich

Dicyclohexyl(4-isopropylphenyl)phosphine

Synonym: Dicyclohexyl[4-(1-methylethyl)phenyl]phosphine

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
InChI Key   NPDGEUFCMFKRER-UHFFFAOYSA-N
mp   24-28 °C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?