• USA Home
  • 711667 - 1,4,7-Tri-Boc-10-(carboxymethyl)-1,4,7,10-tetraazacyclododecane

EMAIL THIS PAGE TO A FRIEND
711667 Sigma-Aldrich

1,4,7-Tri-Boc-10-(carboxymethyl)-1,4,7,10-tetraazacyclododecane

98% (TLC)

Synonym: (4,7,10-Tri-Boc-1,4,7,10-tetraazacyclodecan-1-yl)acetic acid

Purchase

Properties

Related Categories Chelation/Complexation Compounds, Chemical Synthesis, Other Chelation/Complexation Compounds, Synthetic Reagents
assay   ≥97.5% (TLC)
  98% (TLC)
mp   137-142 °C
storage temp.   2-8°C
SMILES string   CC(C)(C)OC(=O)N1CCN(CCN(CCN(CC1)C(=O)OC(C)(C)C)C(=O)OC(C)(C)C)CC(O)=O
InChI   1S/C25H46N4O8/c1-23(2,3)35-20(32)27-12-10-26(18-19(30)31)11-13-28(21(33)36-24(4,5)6)15-17-29(16-14-27)22(34)37-25(7,8)9/h10-18H2,1-9H3,(H,30,31)
InChI key   NDHHFZCDHYVCTI-UHFFFAOYSA-N

Description

Application

Building block used to attach the Cyclen azacrown to a molecular scaffold; selective cleavage of the HIV-1 TAR-RNA with a peptide-cyclen conjugate

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?