• USA Home
  • 712418 - N-(4-Chlorobenzylidene)-4-methylbenzenesulfonamide

EMAIL THIS PAGE TO A FRIEND
712418 Sigma-Aldrich

N-(4-Chlorobenzylidene)-4-methylbenzenesulfonamide

97%

  • CAS Number 3157-65-1

  • Empirical Formula (Hill Notation) C14H12ClNO2S

  • Molecular Weight 293.77

  •  Beilstein/REAXYS Number 2656019

  •  MDL number MFCD02172832

  •  PubChem Substance ID 329763130

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Imines & Amidines, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks,
assay   97%
mp   175-180 °C
SMILES string   Cc1ccc(cc1)S(=O)(=O)\N=C\c2ccc(Cl)cc2
InChI   1S/C14H12ClNO2S/c1-11-2-8-14(9-3-11)19(17,18)16-10-12-4-6-13(15)7-5-12/h2-10H,1H3/b16-10+
InChI key   LKDZCWUUTSGZNH-MHWRWJLKSA-N

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?