• USA Home
  • 712876 - (R)-(−)-2-Methoxy-1-phenylethylamine

EMAIL THIS PAGE TO A FRIEND
712876 Sigma-Aldrich

(R)-(−)-2-Methoxy-1-phenylethylamine

≥95.0% (HPLC)

Synonym: (R)-α-(Methoxymethyl)benzylamine

  • CAS Number 64715-85-1

  • Empirical Formula (Hill Notation) C9H13NO

  • Molecular Weight 151.21

  •  Beilstein/REAXYS Number 4799722

  •  MDL number MFCD01862274

  •  PubChem Substance ID 329763156

  •  NACRES NA.22

Purchase

Properties

Related Categories Amines, Building Blocks, C9, Chemical Synthesis, Ethers,
InChI Key   CMTDMIYJXVBUDX-VIFPVBQESA-N
assay   ≥95.0% (HPLC)
  95.0-105.0% (with HClO4, T)
optical activity   [α]/D -52.0±2.0°, c = 1 in chloroform
storage temp.   2-8°C

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?