• USA Home
  • 712914 - (S)-2-(tert-Butyldiphenylsilanyloxy)propionaldehyde

EMAIL THIS PAGE TO A FRIEND
712914 Sigma-Aldrich

(S)-2-(tert-Butyldiphenylsilanyloxy)propionaldehyde

≥97.0% (GC)

Synonym: (S)-2-(tert-Butyldiphenylsilyloxy)propanal

  • CAS Number 87696-33-1

  • Empirical Formula (Hill Notation) C19H24O2Si

  • Molecular Weight 312.48

  •  Beilstein/REAXYS Number 4192984

  •  MDL number MFCD15143654

  •  PubChem Substance ID 329763159

  •  NACRES NA.22

Purchase

Properties

Related Categories Aldehydes, Asymmetric Synthesis, Building Blocks, C13-C60, Carbonyl Compounds,
Quality Level   100
assay   ≥97.0% (GC)
optical activity   [α]/D -17±3°, c = 1 in chloroform
storage temp.   −20°C
SMILES string   C[C@H](O[Si](c1ccccc1)(c2ccccc2)C(C)(C)C)C=O
InChI   1S/C19H24O2Si/c1-16(15-20)21-22(19(2,3)4,17-11-7-5-8-12-17)18-13-9-6-10-14-18/h5-16H,1-4H3/t16-/m0/s1
InChI key   RRMPJLZPEZKPND-INIZCTEOSA-N

Description

Packaging

500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?