• USA Home
  • 713287 - 1,2-Di-Boc-piperidazine

EMAIL THIS PAGE TO A FRIEND
713287 Sigma-Aldrich

1,2-Di-Boc-piperidazine

≥98.0% (GC)

Synonym: Tetrahydro-pyridazine-1,2-dicarboxylic acid di-tert-butyl ester

  • CAS Number 340256-13-5

  • Empirical Formula (Hill Notation) C14H26N2O4

  • Molecular Weight 286.37

  •  Beilstein/REAXYS Number 9633144

  •  MDL number MFCD19686962

  •  PubChem Substance ID 329763188

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Pyridazines
assay   ≥98.0% (GC)
SMILES string   CC(C)(C)OC(=O)N1CCCCN1C(=O)OC(C)(C)C
InChI   1S/C14H26N2O4/c1-13(2,3)19-11(17)15-9-7-8-10-16(15)12(18)20-14(4,5)6/h7-10H2,1-6H3
InChI key   ONTYDPNTAVCVKA-UHFFFAOYSA-N

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?