• USA Home
  • 714046 - (1S,2S)-trans-2-Amino-1,2,3,4-tetrahydro-1-naphthol hydrochloride

EMAIL THIS PAGE TO A FRIEND
714046 Sigma-Aldrich

(1S,2S)-trans-2-Amino-1,2,3,4-tetrahydro-1-naphthol hydrochloride

≥96.0%

Synonym: (1S,2S)-2-Amino-1-tetralol hydrochloride

  • CAS Number 115563-63-8

  • Empirical Formula (Hill Notation) C10H13NO · HCl

  • Molecular Weight 199.68

  •  Beilstein/REAXYS Number 6577186

  •  MDL number MFCD16621424

  •  PubChem Substance ID 329763244

Purchase

Properties

Related Categories Amino Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   OJKXKJBAGVKYBW-IYPAPVHQSA-N
assay   ≥96.0% (HPLC)
  ≥96.0%
optical purity   enantiomeric excess: ≥97.0%
storage temp.   2-8°C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?