• USA Home
  • 714070 - (1R,2R)-trans-1-Hydroxy-1,2-dihydro-2-naphthyl benzoate

EMAIL THIS PAGE TO A FRIEND
714070 Sigma-Aldrich

(1R,2R)-trans-1-Hydroxy-1,2-dihydro-2-naphthyl benzoate

≥96%

Synonym: (1R,2R)-1,2-Dihydro-1,2-naphthalenediol 2-benzoate

  • CAS Number 337376-51-9

  • Empirical Formula (Hill Notation) C17H14O3

  • Molecular Weight 266.29

  •  Beilstein/REAXYS Number 8843119

  •  MDL number MFCD16621427

  •  PubChem Substance ID 329763249

Purchase

Properties

Related Categories Alcohols, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Esters,
InChI Key   HTGSBYSSVSBBBO-HZPDHXFCSA-N
assay   ≥96%
  ≥96.0% (HPLC)
optical purity   enantiomeric excess: ≥97.0%
storage temp.   2-8°C

Description

Packaging

250 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?