• USA Home
  • 714860 - 1,8-Diazabicyclo[5.4.0]undec-7-ene solution

EMAIL THIS PAGE TO A FRIEND
714860 Sigma-Aldrich

1,8-Diazabicyclo[5.4.0]undec-7-ene solution

1 M in ethyl acetate

Synonym: DBU solution

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Synthetic Reagents More...
InChI Key   GQHTUMJGOHRCHB-UHFFFAOYSA-N
concentration   1 M in ethyl acetate
refractive index   n20/D 1.395
density   0.908 g/mL at 25 °C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1173 - class 3 - PG 2 - Ethyl acetate solution
WGK Germany 
2
Flash Point(F) 
26.6 °F
Flash Point(C) 
-3 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Related Content

Molarity Calculator & Normality Calculator for Acids & Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution. To ...

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?