• USA Home
  • 717053 - cis-4-Fluoro-L-prolinamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
717053 Sigma-Aldrich

cis-4-Fluoro-L-prolinamide hydrochloride

97%

Synonym: (2S,4S)-4-Fluoropyrolidine-2-carboxamide hydrochloride

Purchase

Properties

Related Categories Asymmetric Synthesis, Building Blocks, Chemical Synthesis, Chiral Building Blocks, Fluorinated Building Blocks,
InChI Key   YIEACUBSPQGCLW-MMALYQPHSA-N
assay   97%
optical activity   [α]22/D −45.0°, c = 0.5 in methanol
mp   240-245 °C

Description

Packaging

500 mg in amber glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?