• USA Home
  • 717487 - Potassium 6-fluoropyridine-3-trifluoroborate

EMAIL THIS PAGE TO A FRIEND
717487 Sigma-Aldrich

Potassium 6-fluoropyridine-3-trifluoroborate

95%

  • Empirical Formula (Hill Notation) C5H3BF4KN

  • Molecular Weight 202.99

  •  MDL number MFCD09993004

  •  PubChem Substance ID 329763496

  •  NACRES NA.22

Purchase

Properties

Related Categories Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Trifluoroborate Salts, Organometallic Reagents, Trifluoroborate Salts More...
Quality Level   100
assay   95%
mp   >300 °C
storage temp.   2-8°C
SMILES string   [K+].Fc1ccc(cn1)[B-](F)(F)F
InChI   1S/C5H3BF4N.K/c7-5-2-1-4(3-11-5)6(8,9)10;/h1-3H;/q-1;+1
InChI key   FRIUXLLFAUQFID-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

500 mg in glass insert

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Organotrifluoroborates as Coupling Partners in Suzuki-Miyaura Reactions

Suzuki-Miyaura cross-coupling reactions are some of the most common methods for the formation of C–C bonds in organic chemistry. The use of some boronic acids is complicated by their instability and ...
Josephine Nakhla
Chemfiles Volume 10 Article 3
Keywords: Addition reactions, Chemfiles, Coupling reactions, Cross couplings, Methods, Precipitation

Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?