• USA Home
  • 718807 - 2,6-Dichloro-3-(tributylstannyl)pyrazine

EMAIL THIS PAGE TO A FRIEND
718807 Sigma-Aldrich

2,6-Dichloro-3-(tributylstannyl)pyrazine

97%

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organotin, Organotins
InChI Key   IJOQAXKEUUCPAJ-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.535
density   1.260 g/mL at 25 °C

Description

Packaging

1, 5 g in glass bottle

500 mg in glass bottle

Safety & Documentation

Safety Information

RIDADR 
UN 2788 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles

Articles

Heterocyclic Organotin Reagents for Stille Coupling | Chemfiles Volume 11 Article 1

Stille reactions remain one of the most viable methods for the formation of C–C bonds in organic chemistry.1 Their use has been highlighted in various areas, including countless natural product synth...
Aaron Thornton
Chemfiles Volume 11 Article 1
Keywords: Applications, Cross couplings, Eliminations, Materials Science, Methods, Name reactions, Stille coupling, Stille reaction

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?