• USA Home
  • 719145 - Plexcore® OC RG-1155 organic conductive ink

EMAIL THIS PAGE TO A FRIEND
719145 Sigma-Aldrich

Plexcore® OC RG-1155 organic conductive ink

Synonym: Plexcore® OC organic conductive ink

Purchase

Properties

Related Categories 3D Printing Materials for Research and Development, Bioelectronics, Biosensing and Bioimaging, Conductive Inks, Conductive Inks for 3D Printing,
form   liquid
concentration   2.0%
resistivity   300-1000 Ω-cm
pH   7.8
viscosity   8.6 cP(25 °C)
density   0.938 g/mL at 25 °C
storage temp.   2-8°C

Description

Packaging

25 mL in glass bottle

Warning

Do not freeze. Quality is compromised if ink goes through freeze-thaw cycle even once.

Legal Information

Plexcore is a registered trademark of Plextronics, Inc.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
78.8 °F
Flash Point(C) 
26 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

3D Printing of Carbon Fiber-Reinforced Composites

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries, most typically using a layer-by-layer fabrication process. 3D pri...
Zhenyu Bo* (Ph.D Candidate at Northwestern University) and Jia Choi*, PhD, Product Manager


*Materials Science Product Management Team, MilliporeSigma, Milwaukee, WI.
Keywords: Deposition, Nanomaterials, Nanotubes

Graphene-Based Transparent Conductive Electrodes

Kehan Yu1 and Junhong Chen2* 1Department of Chemical Engineering Case Western Reserve University Cleveland, Ohio 44106 2Department of Mechanical Engineering University of Wisconsin-Milwaukee, Milwauk...
Keywords: Catalysis, Centrifugation, Chemical vapor deposition, Deposition, Electronics, Microelectronics, Nanomaterials, Nanotubes, Oxidations, PAGE, Positron Emission Tomography, Purification, Raman spectroscopy, Reductions, Semiconductor, Solar cells, Solvents, Sonication, Spectroscopy

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?