• USA Home
  • 720275 - Potassium 6-methoxy-3-pyridyltrifluoroborate

EMAIL THIS PAGE TO A FRIEND
720275 Sigma-Aldrich

Potassium 6-methoxy-3-pyridyltrifluoroborate

95%

  • Empirical Formula (Hill Notation) C6H6BF3KNO

  • Molecular Weight 215.02

  •  MDL number MFCD09993111

  •  PubChem Substance ID 329762994

  •  NACRES NA.22

Purchase

Properties

Related Categories Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Trifluoroborate Salts, Organometallic Reagents, Trifluoroborate Salts More...
Quality Level   100
assay   95%
mp   75-80 °C
SMILES string   [K+].COc1ccc(cn1)[B-](F)(F)F
InChI   1S/C6H6BF3NO.K/c1-12-6-3-2-5(4-11-6)7(8,9)10;/h2-4H,1H3;/q-1;+1
InChI key   ASWXYOAUUZSFCK-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

250 mg in glass insert

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?