• USA Home
  • 723975 - Potassium 4-(N,N-dimethylamino)phenyltrifluoroborate

EMAIL THIS PAGE TO A FRIEND
723975 Sigma-Aldrich

Potassium 4-(N,N-dimethylamino)phenyltrifluoroborate

Synonym: Potassium [4-(dimethylamino)phenyl]trifluoroborate

  • CAS Number 1187951-61-6

  • Empirical Formula (Hill Notation) C8H10BF3KN

  • Molecular Weight 227.08

  •  MDL number MFCD09993289

  •  PubChem Substance ID 329764048

  •  NACRES NA.22

Purchase

Properties

Related Categories Aryl Trifluoroborate Salts, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents, Trifluoroborate Salts More...
mp   >300 °C
SMILES string   [K+].CN(C)c1ccc(cc1)[B-](F)(F)F
InChI   1S/C8H10BF3N.K/c1-13(2)8-5-3-7(4-6-8)9(10,11)12;/h3-6H,1-2H3;/q-1;+1
InChI key   RFFDTHVZSLGINK-UHFFFAOYSA-N

Description

Application

Reactant for:
• Suzuki-Miyuara cross-coupling reactions

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?