• USA Home
  • 724130 - (11bS)-4,4-Dibutyl-4,5-dihydro-2,6-bis[3,5-bis(trifluoromethyl)phenyl]-3H-dinaphtho[2,1-c:1′,2′-e]phosphepinium bromide

EMAIL THIS PAGE TO A FRIEND
724130 Sigma-Aldrich

(11bS)-4,4-Dibutyl-4,5-dihydro-2,6-bis[3,5-bis(trifluoromethyl)phenyl]-3H-dinaphtho[2,1-c:1′,2′-e]phosphepinium bromide

95%

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Phase Transfer Reaction
InChI Key   CHNMXCSNZAIEBE-UHFFFAOYSA-M
assay   95%
optical activity   [α]22/D −32.0°, c = 0.5 in chloroform
mp   258-263 °C

Description

Packaging

50 mg in amber glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?