• USA Home
  • 724157 - 4′-Chlorobiphenyl-4-sulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
724157 Sigma-Aldrich

4′-Chlorobiphenyl-4-sulfonyl chloride

97%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonyl Halides, Sulfur Compounds More...
Quality Level   100
assay   97%
mp   108-110 °C
SMILES string   Clc1ccc(cc1)-c2ccc(cc2)S(Cl)(=O)=O
InChI   1S/C12H8Cl2O2S/c13-11-5-1-9(2-6-11)10-3-7-12(8-4-10)17(14,15)16/h1-8H
InChI key   NWYUSJMIHFIMTA-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?