• USA Home
  • 725749 - Potassium octyltrifluoroborate

EMAIL THIS PAGE TO A FRIEND
725749 Sigma-Aldrich

Potassium octyltrifluoroborate

Synonym: Potassium oct-1-yltrifluoroborate, Potassium octane-1-trifluoroborate

Purchase

Properties

Related Categories Alkyl Trifluoroborate Salts, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents, Trifluoroborate Salts More...
mp   74-78 °C
SMILES string   [K+].CCCCCCCC[B-](F)(F)F
InChI   1S/C8H17BF3.K/c1-2-3-4-5-6-7-8-9(10,11)12;/h2-8H2,1H3;/q-1;+1
InChI key   APRQWJZXYZWRAM-UHFFFAOYSA-N

Description

Application

Reactant for:
• Hydrolysis with silica gel and water for preparation of boronic acids and esters
• Suzuki-Miyaura cross-coupling
• Palladium-catalyzed Suzuki-Miyaura-type cross-coupling reaction with alkenyl bromides
• Hydrodeboration reactions

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?