• USA Home
  • 726222 - tert-Butyl N-[(2-methylphenyl)methylene]carbamate

EMAIL THIS PAGE TO A FRIEND
726222 Sigma-Aldrich

tert-Butyl N-[(2-methylphenyl)methylene]carbamate

97%

Synonym: N-(2-Methylphenylmethylene)carbamic acid tert-butyl ester, N-Boc-2-methylbenzylidenamine, o-Tolualdehyde N-(tert-butoxycarbonyl)imine

  • CAS Number 479423-37-5

  • Empirical Formula (Hill Notation) C13H17NO2

  • Molecular Weight 219.28

  •  Beilstein/REAXYS Number 10532568

  •  MDL number MFCD17677172

  •  PubChem Substance ID 329764216

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Imines & Amidines, Nitrogen Compounds - Organic Building Blocks, Organic Building Blocks More...
assay   ≥96.5% (GC)
  97%
storage temp.   room temp
SMILES string   Cc1ccccc1\C=N\C(=O)OC(C)(C)C
InChI   1S/C13H17NO2/c1-10-7-5-6-8-11(10)9-14-12(15)16-13(2,3)4/h5-9H,1-4H3/b14-9+
InChI key   WNCIPIOGTZEGJI-NTEUORMPSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
248 °F
Flash Point(C) 
120 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?