• USA Home
  • 726486 - (S)-(−)-1-(1-Naphthyl)ethylamine

EMAIL THIS PAGE TO A FRIEND
726486 Sigma-Aldrich

(S)-(−)-1-(1-Naphthyl)ethylamine

ChiPros®, produced by BASF, ≥98.5% (GC)

Synonym: (S)-(−)-α-Methyl-1-naphthalenemethylamine

  • CAS Number 10420-89-0

  • Linear Formula C10H7CH(CH3)NH2

  • Molecular Weight 171.24

  •  Beilstein/REAXYS Number 2208024

  •  MDL number MFCD00064179

  •  PubChem Substance ID 329764239

Purchase

Properties

Related Categories Amines, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
grade   produced by BASF
InChI Key   RTCUCQWIICFPOD-VIFPVBQESA-N
assay   ≥98.5% (GC)
optical purity   enantiomeric excess: ≥98.5%
refractive index   n20/D 1.623 (lit.)
bp   153 °C/11 mmHg (lit.)
density   1.067 g/mL at 20 °C (lit.)

Description

Packaging

5, 25 g in glass bottle

Legal Information

ChiPros is a registered trademark of BASF SE

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles

Articles

ChiPros Chiral Amines

Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries. While classically available through racem...
Daniel Weibel
Chemfiles Volume 11 Article 1
Keywords: Building blocks, Catalysis, Chiral auxiliaries, Organic synthesis, Resolving agents

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?