• USA Home
  • 727458 - 2-Azidobenzoic acid solution

EMAIL THIS PAGE TO A FRIEND
727458 Sigma-Aldrich

2-Azidobenzoic acid solution

~0.25 M in tert-butyl methyl ether, ≥95.0% (HPLC)

Synonym: o-Azidobenzoic acid solution

  • CAS Number 31162-13-7

  • Empirical Formula (Hill Notation) C7H5N3O2

  • Molecular Weight 163.13

  •  Beilstein/REAXYS Number 1950619

  •  MDL number MFCD00211421

  •  PubChem Substance ID 329764320

  •  NACRES NA.22

Purchase

Properties

Related Categories Aromatic Azides, Azides, Building Blocks, C7, Carbonyl Compounds,
assay   ≥95.0% (HPLC)
concentration   ~0.25 M in tert-butyl methyl ether
impurities   ≤2.0% water
storage temp.   −20°C
SMILES string   OC(=O)c1ccccc1N=[N+]=[N-]
InChI   1S/C7H5N3O2/c8-10-9-6-4-2-1-3-5(6)7(11)12/h1-4H,(H,11,12)
InChI key   JCBWQNLTYXTHBZ-UHFFFAOYSA-N

Description

Packaging

10 mL in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2398 3 / PGII
WGK Germany 
3
Flash Point(F) 
-27.4 °F
Flash Point(C) 
-33 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?