• USA Home
  • 72829 - (S)-3-(Fmoc-amino)-4-(2-naphthyl)butyric acid

EMAIL THIS PAGE TO A FRIEND
72829 Sigma-Aldrich

(S)-3-(Fmoc-amino)-4-(2-naphthyl)butyric acid

≥95%

Synonym: Fmoc-β-2-Homonal-OH, Fmoc-4-(2-naphthyl)-L-β-homoalanine

Purchase

Properties

Related Categories Beta-Amino Acids, Chemical Biology, Chemical Synthesis, Peptide Synthesis and Peptide Chemistry, Unnatural Amino Acids & Derivatives More...
assay   ≥95%
application(s)   peptide synthesis: suitable
functional group   Fmoc
storage temp.   2-8°C
SMILES string   OC(=O)C[C@H](Cc1ccc2ccccc2c1)NC(=O)OCC3c4ccccc4-c5ccccc35
InChI   1S/C29H25NO4/c31-28(32)17-22(16-19-13-14-20-7-1-2-8-21(20)15-19)30-29(33)34-18-27-25-11-5-3-9-23(25)24-10-4-6-12-26(24)27/h1-15,22,27H,16-18H2,(H,30,33)(H,31,32)/t22-/m0/s1
InChI key   LQGDRWACMZMSCD-QFIPXVFZSA-N

Description

Packaging

500 mg in poly tube

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

β-Amino Acids and Homologs

Although they are less abundant than their α-analogues, β-amino acids occur in nature both in free form and bound to peptides. Oligomers composed exclusively of β-amino acids (so-called β-peptides) m...
Matthias Junkers
Aldrich ChemFiles 2008, 8.7, 11.
Keywords: Chemfiles, Degradations, Metabolism, Peptidomimetics

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?