• USA Home
  • 728861 - Potassium 2,4-bis(trifluoromethyl)phenyltrifluoroborate

EMAIL THIS PAGE TO A FRIEND
728861 Sigma-Aldrich

Potassium 2,4-bis(trifluoromethyl)phenyltrifluoroborate

95%

Purchase

Properties

Related Categories Aryl Trifluoroborate Salts, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents, Trifluoroborate Salts More...
assay   95%
mp   173-178 °C
storage temp.   2-8°C
SMILES string   [K+].F[B-](F)(F)c1ccc(cc1C(F)(F)F)C(F)(F)F
InChI   1S/C8H3BF9.K/c10-7(11,12)4-1-2-6(9(16,17)18)5(3-4)8(13,14)15;/h1-3H;/q-1;+1
InChI key   OXVQSBLFWHNZJA-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?