• USA Home
  • 731846 - CdSe/ZnS quantum dots

EMAIL THIS PAGE TO A FRIEND
731846 Sigma-Aldrich

CdSe/ZnS quantum dots

fluorescence λem 590 nm, 5 mg/mL in toluene/aliphatic amine

Synonym: NanoOrange CdSe/ZnS - 590

  • Linear Formula CdSe/ZnS

Purchase

Properties

Related Categories Core Type Quantum Dots, Core-Shell Type Quantum Dots, Materials Science, Nanomaterials, Quantum Dots More...
form   liquid
availability   available only in USA
concentration   5 mg/mL in toluene/aliphatic amine
color   orange
density   0.866 g/mL at 20 °C (lit.)
fluorescence   λem 590 nm
  , FWHM <60 nm, quantum yield >80%
storage temp.   2-8°C

Description

Legal Information

Product of Nanosys, Inc. For research and development only; excluding use for biological assay systems and/or combinatorial chemistry

NanoOrange is a trademark of Nanosys, Inc.

Safety & Documentation

Safety Information

Signal word 
Danger
Target organs 
Central nervous system
RIDADR 
UN 2924 8(3) / PGII
WGK Germany 
WGK 3
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Nanostructured Materials Through Ultrasonic Spray Pyrolysis | Technical Article

Jin Ho Bang,1,2 Yuri T. Didenko,3 Richard J. Helmich,1,4 and Kenneth S. Suslick1* 1School of Chemical Sciences, University of Illinois at Urbana-Champaign, 600 S. Mathews Av., Urbana, IL 61801 USA 2C...
Keywords: Antimicrobials, Cancer, Catalysis, Characterizations, Chemical reactions, Condensations, Deposition, Diffusion, Electronics, Evaporation, Flow cytometry, Nanomaterials, Oxidations, Photovoltaics, Precipitation, Purification, Reductions, Semiconductor, Separation, Solar cells, Solvents, Thin film deposition

Nanostructured Materials Through Ultrasonic Spray Pyrolysis

Advances in materials have often been led by the development of new synthetic methods that provide control over size, morphology and structure. The preparation of materials in a scalable and continuo...
Jin Ho Bang, Yuri T. Didenko, Richard J. Helmich, and Kenneth S. Suslick
Material Matters, Volume 7, Number 2, 15–20
Keywords: Antimicrobials, Cancer, Catalysis, Characterizations, Chemical reactions, Condensations, Deposition, Diffusion, Electronics, Evaporation, Flow cytometry, Nanomaterials, Oxidations, Photovoltaics, Precipitation, Purification, Reductions, Semiconductor, Separation, Solar cells, Solvents, Thin film deposition

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?