• USA Home
  • 73464 - (RP,R′P)-1,1′-Bis[(S)-α-(dimethylamino)benzyl]-2,2′-bis(diphenylphosphino)ferrocene

EMAIL THIS PAGE TO A FRIEND
73464 Sigma-Aldrich

(RP,R′P)-1,1′-Bis[(S)-α-(dimethylamino)benzyl]-2,2′-bis(diphenylphosphino)ferrocene

≥97%

Synonym: (2S,2′S)-1,1′-Bis[(S)-(dimethylamino)phenylmethyl]-2,2′-bis(diphenylphosphino)ferrocene (acc to CAS), Mandyphos SL-M001-2

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Mandyphos Ligands, Privileged Ligands and Complexes,
InChI Key   HZATWRYPFZEGHM-FKMBXMNYSA-N
assay   ≥97%
optical purity   ee: ≥99%

Description

General description

sold in collaboration with Solvias AG

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?