• USA Home
  • 73472 - (RP,R′P)-1,1′-Bis[(S)-α-(dimethylamino)benzyl]-2,2′-bis[di(3,5-xylyl)phosphino]ferrocene

EMAIL THIS PAGE TO A FRIEND
73472 Sigma-Aldrich

(RP,R′P)-1,1′-Bis[(S)-α-(dimethylamino)benzyl]-2,2′-bis[di(3,5-xylyl)phosphino]ferrocene

≥97%

Synonym: (1S,1′S)- 1,1′-Bis[bis(3,5-dimethylphenyl)phosphino]-2,2′-bis[(S)-(dimethylamino)phenylmethyl]ferrocene (acc to CAS), Mandyphos SL-M009-2

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Mandyphos Ligands, Privileged Ligands and Complexes,
assay   ≥97%
optical purity   ee: ≥99%
SMILES string   [Fe].CN(C)[C@H]([C]1[CH][CH][CH][C]1P(c2cc(C)cc(C)c2)c3cc(C)cc(C)c3)c4ccccc4.CN(C)[C@H]([C]5[CH][CH][CH][C]5P(c6cc(C)cc(C)c6)c7cc(C)cc(C)c7)c8ccccc8
InChI   1S/2C30H33NP.Fe/c2*1-21-15-22(2)18-26(17-21)32(27-19-23(3)16-24(4)20-27)29-14-10-13-28(29)30(31(5)6)25-11-8-7-9-12-25;/h2*7-20,30H,1-6H3;/t2*30-;/m00./s1
InChI key   YUERARBCLXUXCP-RELFUISISA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

1, 5 g in glass bottle

100, 500 mg in glass bottle

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?