• USA Home
  • 735035 - Potassium 2-chlorothiophene-5-trifluoroborate

EMAIL THIS PAGE TO A FRIEND
735035 Sigma-Aldrich

Potassium 2-chlorothiophene-5-trifluoroborate

Purchase

Properties

Related Categories Boronic Acids and Derivatives, Chemical Synthesis, Heteroaryl Trifluoroborate Salts, Organometallic Reagents, Trifluoroborate Salts More...
mp   170-175 °C
SMILES string   [K+].F[B-](F)(F)c1ccc(Cl)s1
InChI   1S/C4H2BClF3S.K/c6-4-2-1-3(10-4)5(7,8)9;/h1-2H;/q-1;+1
InChI key   CBFRQOVNXITSBL-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?